Tlačiť

Anti-CCNT1 Rabbit polyclonal antibody

ABCAAB106968-400
Anti-CCNT1 Rabbit polyclonal antibody
Type: Primary
Antigen: CCNT1 (cyclin T1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR