Tlačiť
Anti-IgG Goat Polyclonal Antibody (Biotin)
Dodávateľ: Biotium
20187.EA 258.05 EUR
20187. BTIU20187-1
Anti-IgG Goat Polyclonal Antibody (Biotin)
Protilátky
This conjugate is prepared by labeling F(ab’)2 fragment of goat anti-mouse IgG (H+L) with biotin.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse