Tlačiť
Anti-IgG2b Goat Polyclonal Antibody (CF™ 594)
Dodávateľ: Biotium
20269-1EA 180.99 EUR
20269-1 20269.
Anti-IgG2b Goat Polyclonal Antibody (CF™ 594)
Protilátky
Type: Secondary
Antigen: IgG2b (Mouse) (mouse IgG2b)
Clonality: polyclonal
Clone:
Conjugation: CF® 594 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse