Tlačiť
Anti-Cripto Rabbit Polyclonal Antibody
Dodávateľ: Thermo Fisher Scientific
PIEAPA116533EA 0 EUR
PIEAPA116533
Anti-Cripto Rabbit Polyclonal Antibody
Protilátky
This antibody is predicted to react with mouse based on 93% sequence homology. The cripto gene is expressed both in ES cells and during the early phases of embryo development, while, in the adult, it is reactivated in a wide range of epithelial cancers.
Type: Primary
Antigen: Cripto
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human