Chcete sa opäť prihlásiť?
Anti-Cripto Rabbit Polyclonal Antibody
Dodávateľ: Thermo Scientific
In Western blot a band is seen at ~18 kDa representing the core form of Cripto1, and at ~36 kDa reporesenting the glycosylated/ post-translationally modified form. Suggested positive control: MDA-MB231 (human breast cancer) lysates. The cripto gene is expressed both in ES cells and during the early phases of embryo development, while, in the adult, it is reactivated in a wide range of epithelial cancers.
Type: Primary
Antigen: Cripto
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Veľkosť Pk Číslo v katalógu VWR Množstvo
100 µL 100 µl PIEAPA116534 Blocked
Blocked Vaša organizácia zablokovala tento produkt. Prosím kontaktujte oddelenie nákupu pre viac informácii.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
ŠPECIFIKÁCIE
Antigen symbol Cripto
Antigen name Teratocarcinoma-derived growth factor 1
Reactivity Mouse, Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoFluorescence Yes
Flow cytometry Yes
Antigen synonyms CRGF, Cripto-3, Cripto-1 Growth Factor, TDGF1, TDGF2, Teratocarcinoma-derived growth factor 1
Gene ID 6997, 21667
Concentration 1.0 mg/ml
Immunogen A synthetic peptide made to an N-terminal portion of the murine cripto protein sequence (between residues 1-50).
Purification Antigen affinity chromatography
Form Liquid
Storage buffer tris glycine, 0.15M NaCl, 0.05% sodium azide
Storage temperature Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Shipping temperature Ice
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo