Tlačiť
Anti-ATRIP Rabbit Polyclonal Antibody
Dodávateľ: Thermo Fisher Scientific
PIEAPA116582EA 0 EUR
PIEAPA116582
Anti-ATRIP Rabbit Polyclonal Antibody
Protilátky
Suggested positive control: Hela whole cell extract, antigen standard for ATRIP (transient overexpression lysate). ATRIP is an 86 kDa protein that is phosphorylated by ATR, regulates ATR expression, and is an essential component of the DNA damage checkpoint pathway. It localizes to the intranuclear foci after DNA damage or inhibition of replication.
Type: Primary
Antigen: ATRIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human