Tlačiť

Anti-p75NTR / CD271 Goat Polyclonal Antibody

Dodávateľ: Everest Biotechnology
EVBIEB11125EA 361.99 EUR
EVBIEB11125
Anti-p75NTR / CD271 Goat Polyclonal Antibody
Protilátky
Type: Primary
Antigen: p75NTR / CD271
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR