Tlačiť
Anti-PRKCG Rabbit Polyclonal Antibody
Dodávateľ: GenScript Biotech
GENSA00904-40EA 212.63 EUR
GENSA00904-40
Anti-PRKCG Rabbit Polyclonal Antibody
Type: Primary
Antigen: PRKCG (protein kinase C, gamma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Order Now