Tlačiť
Anti-CXCL10 Mouse Monoclonal Antibody [clone: 1G12E6]
Dodávateľ: GenScript Biotech
GENSA01561-40EA 189.87 EUR
GENSA01561-40
Anti-CXCL10 Mouse Monoclonal Antibody [clone: 1G12E6]
Type: Primary
Antigen: CXCL10 (chemokine (C-X-C motif) ligand 10)
Clonality: monoclonal
Clone: 1G12E6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity:
Order Now