Tlačiť
Anti-alpha 1 Glycine Receptor Rabbit Polyclonal Antibody
Dodávateľ: GenScript Biotech
GENSA01636-40EA 189.87 EUR
GENSA01636-40
Anti-alpha 1 Glycine Receptor Rabbit Polyclonal Antibody
Type: Primary
Antigen: alpha 1 Glycine Receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Order Now