Tlačiť

Anti-IgG F(ab')2 Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Dodávateľ: Rockland Immunochemicals
ROCK712-1231EA 536.26 EUR
ROCK712-1231
Anti-IgG F(ab')2 Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Protilátky
Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring extremely low background levels, absence of F(c) mediated binding, lot-to-lot consistency, high titer and specificity.

Recommended Dilutions: FLISA: 1:50 - 1:200; Immunofluorescence: 1:50 - 1:200; Flow Cytometry: 1:500 - 1:2500; Western Blot: 1:50 - 1:200
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope: F(ab')2
Host: Goat
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR