Tlačiť

Anti-HNRNPDL Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Dodávateľ: US Biological
USBI036670-APCEA 949.22 EUR
USBI036670-APC
Anti-HNRNPDL Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Protilátky
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: HNRNPDL (heterogeneous nuclear ribonucleoprotein D-like)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR