Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

Anti-HNRNPDL Rabbit Polyclonal Antibody (Biotin)

Dodávateľ: US Biological
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: HNRNPDL (heterogeneous nuclear ribonucleoprotein D-like)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human



Log in to see your contract pricing and availability.
  Zapamätať si ma na tomto zariadení
 

OBJEDNAŤ

Rozšíriť všetko / Schovať všetko
Veľkosť Pk Číslo v katalógu VWR Množstvo
200 µl 200 µl USBI036670-BIOTIN
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ŠPECIFIKÁCIE

Antigen symbol HNRNPDL
Antigen name heterogeneous nuclear ribonucleoprotein D-like
Reactivity Human
Conjugation Biotin
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Amino acid number 214 to 244
Epitope internal
Purification purified by Protein A chromatography
Form liquid
Storage buffer PBS, pH 7,2
Storage temperature Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo