Tlačiť

Anti-HNRNPLL Rabbit Polyclonal Antibody

Dodávateľ: US Biological
USBI036672EA 654.02 EUR
USBI036672
Anti-HNRNPLL Rabbit Polyclonal Antibody
Protilátky
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: HNRNPLL (heterogeneous nuclear ribonucleoprotein L-like)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR