Tlačiť

Anti-HOMEZ Rabbit Polyclonal Antibody (Biotin)

Dodávateľ: US Biological
USBI036674-BIOTINEA 947.16 EUR
USBI036674-BIOTIN
Anti-HOMEZ Rabbit Polyclonal Antibody (Biotin)
Protilátky
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50

Type: Primary
Antigen: HOMEZ (homeobox and leucine zipper encoding)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR