Tlačiť
Anti-HOOK1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Dodávateľ: US Biological
USBI036675-APCEA 947.16 EUR
USBI036675-APC
Anti-HOOK1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Protilátky
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: HOOK1 (hook microtubule-tethering protein 1)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human