Chcete sa opäť prihlásiť?
Anti-HOPX Rabbit Polyclonal Antibody
Dodávateľ: US Biological
Recommended dilution: Western Blot: 1:1000; Flow Cytometry: 1:10-1:50
Type: Primary
Antigen: HOPX (HOP homeobox)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Veľkosť Pk Číslo v katalógu VWR Množstvo
200 µl 200 µl USBI036677
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
ŠPECIFIKÁCIE
Antigen symbol HOPX
Antigen name HOP homeobox
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Flow cytometry Yes
Epitope C-terminal
Purification purified by Protein A chromatography
Form liquid
Storage buffer PBS, pH 7,2, 0,09% sodium azide
Storage temperature Cool
Shipping temperature Ice
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo