Tlačiť

Anti-HOPX Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Dodávateľ: US Biological
USBI036677-FITCEA 949.22 EUR
USBI036677-FITC
Anti-HOPX Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Protilátky
Type: Primary
Antigen: HOPX (HOP homeobox)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR