Tlačiť

Anti-HOXA1 Rabbit Polyclonal Antibody (Biotin)

Dodávateľ: US Biological
USBI036679-BIOTINEA 947.16 EUR
USBI036679-BIOTIN
Anti-HOXA1 Rabbit Polyclonal Antibody (Biotin)
Protilátky
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: HOXA1 (homeobox A1)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR