Tlačiť
Anti-HOXA1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Dodávateľ: US Biological
USBI036680-APEA 949.22 EUR
USBI036680-AP
Anti-HOXA1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Protilátky
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: HOXA1 (homeobox A1)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse