Tlačiť
Anti-HOXA10 Rabbit Polyclonal Antibody (Biotin)
Dodávateľ: US Biological
USBI036682-BIOTINEA 947.16 EUR
USBI036682-BIOTIN
Anti-HOXA10 Rabbit Polyclonal Antibody (Biotin)
Protilátky
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunofluorescence: 1:10-50
Type: Primary
Antigen: HOXA10 (homeobox A10)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human