Tlačiť

Anti-HOXA13 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Dodávateľ: US Biological
USBI036684-PEEA 949.22 EUR
USBI036684-PE
Anti-HOXA13 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Protilátky
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: HOXA13 (homeobox A13)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR