Tlačiť

Anti-HOXA2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodávateľ: US Biological
USBI036685-HRPEA 947.16 EUR
USBI036685-HRP
Anti-HOXA2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: HOXA2 (homeobox A2)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR