Tlačiť

Anti-HOXA3 Rabbit Polyclonal Antibody (Biotin)

Dodávateľ: US Biological
USBI036686-BIOTINEA 949.22 EUR
USBI036686-BIOTIN
Anti-HOXA3 Rabbit Polyclonal Antibody (Biotin)
Protilátky
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: HOXA3 (homeobox A3)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR