Tlačiť
Anti-NDST4 Rabbit Polyclonal Antibody (Biotin)
Dodávateľ: US Biological
USBI038852-BIOTINEA 949.22 EUR
USBI038852-BIOTIN
Anti-NDST4 Rabbit Polyclonal Antibody (Biotin)
Protilátky
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: NDST4 (N-deacetylase/N-sulfotransferase (heparan glucosaminyl) 4)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human