Tlačiť
Anti-SNX13 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Dodávateľ: United States Biological
USBI042109-APEA 947.16 EUR
USBI042109-AP
Anti-SNX13 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Protilátky
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: SNX13 (sorting nexin 13)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human