Tlačiť
Anti-SNX14 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Dodávateľ: United States Biological
USBI042110-PEEA 947.16 EUR
USBI042110-PE
Anti-SNX14 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Protilátky
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: SNX14 (sorting nexin 14)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse