Tlačiť

Anti-SNX29 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Dodávateľ: United States Biological
USBI042120-APCEA 947.16 EUR
USBI042120-APC
Anti-SNX29 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Protilátky
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!

Type: Primary
Antigen: SNX29 (sorting nexin 29)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR