Tlačiť
Anti-SNX3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Dodávateľ: United States Biological
USBI042121-FITCEA 947.16 EUR
USBI042121-FITC
Anti-SNX3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Protilátky
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: SNX3 (sorting nexin 3)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human