Tlačiť

Anti-SOCS6 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodávateľ: United States Biological
USBI042133-HRPEA 949.22 EUR
USBI042133-HRP
Anti-SOCS6 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50

Type: Primary
Antigen: SOCS6 (suppressor of cytokine signaling 6)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR