Tlačiť

Anti-SOCS7 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Dodávateľ: United States Biological
USBI042134-APEA 947.16 EUR
USBI042134-AP
Anti-SOCS7 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Protilátky
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: SOCS7 (suppressor of cytokine signaling 7)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR