Tlačiť

Anti-SOHLH2 Rabbit Polyclonal Antibody (Biotin)

Dodávateľ: United States Biological
USBI042136-BIOTINEA 949.22 EUR
USBI042136-BIOTIN
Anti-SOHLH2 Rabbit Polyclonal Antibody (Biotin)
Protilátky
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: SOHLH2 (spermatogenesis and oogenesis specific basic helix-loop-helix 2)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR