Tlačiť

Anti-SORBS1 Rabbit Polyclonal Antibody

Dodávateľ: United States Biological
USBI042137EA 652.6 EUR
USBI042137
Anti-SORBS1 Rabbit Polyclonal Antibody
Protilátky
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunofluorescence: 1:10-50

Type: Primary
Antigen: SORBS1 (sorbin and SH3 domain containing 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Hamster, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR