Tlačiť
Anti-WWTR1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Dodávateľ: United States Biological
USBI043909-FITCEA 947.16 EUR
USBI043909-FITC
Anti-WWTR1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Protilátky
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: WWTR1 (WW domain containing transcription regulator 1)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human