Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

Anti-Endothelin Sheep polyclonal antibody unconjugated

Dodávateľ: US Biological
Recommended dilution: ELISA: 1:1000; Immunochemistry: 1:10

Type: Primary
Antigen: Endothelin
Clonality: polyclonal
Conjugation: unconjugated
Host: Sheep
Isotype: IgG
Reactivity: Human

Log in to see your contract pricing and availability.
  Zapamätať si ma na tomto zariadení


Rozšíriť všetko / Schovať všetko
Veľkosť Pk Číslo v katalógu VWR Množstvo
20 µg 20 µG USBIE3098-01D
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen name Endothelin
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Sheep
Isotype IgG
Concentration ~1 mg/ml
Purification purified by immunoaffinity chromatography
Form liquid
Storage buffer 0,05M borate buffer pH 8.5.
Storage temperature Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo