Tlačiť

Anti-Endothelin, pan Mouse monoclonal antibody [clone: 8k43]

USBIE3098-01P
Anti-Endothelin, pan Mouse monoclonal antibody [clone: 8k43]
Type: Primary
Antigen: Endothelin, pan
Clonality: monoclonal
Clone: 8k43
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR