Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

Anti-EDN1 Rabbit polyclonal antibody unconjugated

Dodávateľ: US Biological
Recommended dilution: Immunochemistry: 1:500

Type: Primary
Antigen: EDN1 (endothelin 1)
Clonality: polyclonal
Conjugation: unconjugated
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Log in to see your contract pricing and availability.
  Zapamätať si ma na tomto zariadení


Rozšíriť všetko / Schovať všetko
Veľkosť Pk Číslo v katalógu VWR Množstvo
100 µl 100 µl USBIE3098-24A
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol EDN1
Antigen name endothelin 1
Reactivity Rat, Human, Mouse
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Amino acid number 109 to 125
Format Serum
Form lyophilized powder
Storage temperature Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo