Chcete sa opäť prihlásiť?
Anti-EDN1 Rabbit polyclonal antibody unconjugated
Dodávateľ: US Biological
Recommended dilution: Immunofluorescence: 1:200; Immunochemistry: 1:200
Type: Primary
Antigen: EDN1 (endothelin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Dog, Human, Mouse, Pig, Rabbit, Rat
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Veľkosť Pk Číslo v katalógu VWR Množstvo
50 µl 50 µl USBIE3098-24F
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
ŠPECIFIKÁCIE
Antigen symbol EDN1
Antigen name endothelin 1
Reactivity Pig, Rabbit, Dog, Human, Mouse, Bovine, Rat
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
ImmunoFluorescence Yes
Format Serum
Form lyophilized powder
Storage temperature Cool
Shipping temperature Ice
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo