Tlačiť

Anti-EphA2 Receptor Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodávateľ: United States Biological
USBIE3360-02E-HRPEA 947.16 EUR
USBIE3360-02E-HRP
Anti-EphA2 Receptor Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Recommended dilution: Western Blot: 1:100-1:500; ELISA: 1:1000; Immunochemistry: 1:50-1:100; Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: EphA2 Receptor
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Hamster, Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR