Tlačiť

Anti-EphA4 Receptor Mouse Monoclonal Antibody [clone: 6A70]

Dodávateľ: United States Biological
USBIE3362-01DEA 728.71 EUR
USBIE3362-01D
Anti-EphA4 Receptor Mouse Monoclonal Antibody [clone: 6A70]
Protilátky
Recommended dilution: Western Blot: 1-3?g/ml; Immunoprecipitation: 5 ?g; Immunochemistry: 3-10 ?g/ml

Type: Primary
Antigen: EphA4 Receptor
Clonality: monoclonal
Clone: 6A70
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR