Tlačiť

Anti-EPHA4 Rabbit Polyclonal Antibody (Biotin)

Dodávateľ: United States Biological
USBIE3362-01L-BIOTEA 947.16 EUR
USBIE3362-01L-BIOT
Anti-EPHA4 Rabbit Polyclonal Antibody (Biotin)
Protilátky
Type: Primary
Antigen: EPHA4 (EPH receptor A4)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR