Tlačiť

Anti-EPHA4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodávateľ: United States Biological
USBIE3362-01L-HRPEA 947.16 EUR
USBIE3362-01L-HRP
Anti-EPHA4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: EPHA4 (EPH receptor A4)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR