Tlačiť

Anti-EFNA4 Rabbit polyclonal antibody

USBIE3362-01P
Anti-EFNA4 Rabbit polyclonal antibody
Recommended dilution: Immunochemistry: 1:100

Type: Primary
Antigen: EFNA4 (ephrin-A4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR