Tlačiť

Anti-EPHA4 Rabbit polyclonal antibody

USBIE3362-01R
Anti-EPHA4 Rabbit polyclonal antibody
Type: Primary
Antigen: EPHA4 (EPH receptor A4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR