Tlačiť

Anti-EphA4b Rabbit polyclonal antibody

USBIE3362-03
Anti-EphA4b Rabbit polyclonal antibody
Recommended dilution: ELISA: 1:5000-1:20000

Type: Primary
Antigen: EphA4b
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR