Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

Anti-EPHA5 Mouse monoclonal antibody FITC (Fluorescein) [clone: 10i34]

Dodávateľ: US Biological
Recommended dilution: FLISA: 1:4000; Western Blot: 1:2000
Do not freeze!

Type: Primary
Antigen: EPHA5 (EPH receptor A5)
Clonality: monoclonal
Clone: 10i34
Conjugation: FITC (Fluorescein)
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human

Log in to see your contract pricing and availability.
  Zapamätať si ma na tomto zariadení


Rozšíriť všetko / Schovať všetko
Veľkosť Pk Číslo v katalógu VWR Množstvo
100 µl 100 µl USBIE3363-01A-FITC
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol EPHA5
Antigen name EPH receptor A5
Reactivity Human
Conjugation FITC (Fluorescein Isothiocyanate)
Clonality Monoclonal
Antibody type Primary
Clone 10i34
Host Mouse
Isotype IgG1 kappa
Western blot Yes
Purification purified by Protein G chromatography
Form liquid
Storage buffer PBS, pH 7,2, contains ~0,09% sodium azide
Storage temperature Cool
Shipping temperature Ice
Tested applications Fluoresence ELISA
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo