Tlačiť

Anti-EPHA5 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 10i34]

Dodávateľ: United States Biological
USBIE3363-01A-HRPEA 842.19 EUR
USBIE3363-01A-HRP
Anti-EPHA5 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 10i34]
Recommended dilution: ELISA: 1:4000; Western Blot: 1:2000

Type: Primary
Antigen: EPHA5 (EPH receptor A5)
Clonality: monoclonal
Clone: 10i34
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR