Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

Anti-EFNA5 Rabbit polyclonal antibody unconjugated

Dodávateľ: US Biological
Recommended dilution: Immunofluorescence: 5 μg/ml; Western Blot: 1-3μg/ml; Immunoprecipitation: 5-10 μg/ Immunoprecipitation: reaction

Type: Primary
Antigen: EFNA5 (ephrin-A5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse




 

OBJEDNAŤ

Rozšíriť všetko / Schovať všetko
Veľkosť Pk Číslo v katalógu VWR Množstvo
100 µg 100 µG USBIE3363-01C-100
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ŠPECIFIKÁCIE

Antigen symbol EFNA5
Antigen name ephrin-A5
Reactivity Mouse
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Concentration ~0,125 mg/ml
Purification purified by immunoaffinity chromatography
Form liquid
Storage buffer PBS, pH 7,4, 0,1% sodium azide. before the addition of glycerol to 40%
Storage temperature Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo