Tlačiť
Anti-EPHA5 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Dodávateľ: United States Biological
USBIE3363-01D-APEA 947.16 EUR
USBIE3363-01D-AP
Anti-EPHA5 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Protilátky
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!
Type: Primary
Antigen: EPHA5 (EPH receptor A5)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse