Tlačiť

Anti-EPHA5 Rabbit Polyclonal Antibody

Dodávateľ: United States Biological
USBIE3363-01FEA 652.6 EUR
USBIE3363-01F
Anti-EPHA5 Rabbit Polyclonal Antibody
Protilátky
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: EPHA5 (EPH receptor A5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR